4-Bromo-3-chloro-2-fluoropyridine

Lieferant: Apollo Scientific

PC904659-5G PC904659-25G
APOSPC904659-5GEA 139 EUR
APOSPC904659-5G APOSPC904659-25G
4-Bromo-3-chloro-2-fluoropyridine
4-Bromo-3-chloro-2-fluoropyridine
Formel: C₅H₂BrClFN
Molecular Weight: 210.43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040607
CAS-Nummer: 1017793-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR