Methyl-2-(2-fluorophenyl)acetate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-fluorophenylacetate, Methyl 2-(2-fluorophenyl)acetate

PC904715-25G
APOSPC904715-25GEA 68.3 EUR
APOSPC904715-25G
Methyl-2-(2-fluorophenyl)acetate 98%
Methyl-2-(2-fluorophenyl)acetate
Formel: C₉H₉FO₂
Molecular Weight: 168.167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800605
CAS-Nummer: 57486-67-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR