4-Amino-5-(difluoromethyl)-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific
Achtung

PC904794-1G PC904794-5G
APOSPC904794-1GEA 66.7 EUR
APOSPC904794-1G APOSPC904794-5G
4-Amino-5-(difluoromethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(difluoromethyl)-4H-1,2,4-triazole-3-thiol
Formel: C₃H₄F₂N₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD03423399
CAS-Nummer: 832740-00-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR