3-(4-Fluorphenyl)isoxazol 95%
Lieferant: Apollo Scientific
PC904842-1G
PC904842-250MG
APOSPC904842-1GEA
273
EUR
APOSPC904842-1G
APOSPC904842-250MG
3-(4-Fluorphenyl)isoxazol 95%
3-(4-Fluorophenyl)isoxazole
Formel:
C₉H₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08446385 CAS-Nummer: 651059-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...