1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid 95%
Lieferant: Apollo Scientific
PC904931-1G
PC904931-5G
APOSPC904931-1GEA
132
EUR
APOSPC904931-1G
APOSPC904931-5G
1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid 95%
1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid
Formel:
C₆H₆F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22421758 CAS-Nummer: 1001026-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...