5-Fluoro-3-methyl-1H-indole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

PC905108-1G PC905108-5G
APOSPC905108-1GEA 139 EUR
APOSPC905108-1G APOSPC905108-5G
5-Fluoro-3-methyl-1H-indole-2-carbaldehyde 95%
5-Fluoro-3-methyl-1H-indole-2-carbaldehyde
Formel: C₁₀H₈FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD06011083
CAS-Nummer: 842972-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR