2-(2-Chloro-3-fluorophenyl)acetic acid 95%

Lieferant: Apollo Scientific

PC905262-5G
APOSPC905262-5GEA 85.7 EUR
APOSPC905262-5G
2-(2-Chloro-3-fluorophenyl)acetic acid 95%
2-(2-Chloro-3-fluorophenyl)acetic acid
Formel: C₈H₆ClFO₂
Molecular Weight: 188,5800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09925136
CAS-Nummer: 1000523-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR