3,5,6-Trifluor-2-pyridinol 95%

Lieferant: Apollo Scientific

Synonyme: 3,5,6-Trifluoro-2-pyridinol, 3,5,6-Trifluoro-2-pyridone

PC905307-1G PC905307-5G
APOSPC905307-1GEA 190 EUR
APOSPC905307-1G APOSPC905307-5G
3,5,6-Trifluor-2-pyridinol 95%
3,5,6-Trifluor-2-pyridinol
Formel: C₅H₂F₃NO
Molecular Weight: 149,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114299
CAS-Nummer: 75777-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR