Fluoromethyl phenyl sulphone 98%

Lieferant: Apollo Scientific

Synonyme: Fluoromethylsulphonylbenzene, ((Fluoromethyl)sulphonyl)benzene

PC905535-5G
APOSPC905535-5GEA 144 EUR
APOSPC905535-5G
Fluoromethyl phenyl sulphone 98%
Fluoromethyl phenyl sulphone
Formel: C₇H₇FO₂S
Molecular Weight: 174,19 g/mol
Schmelzpunkt: 53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191650
CAS-Nummer: 20808-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR