Tetrabutylammonium difluorotriphenylsilicate 97%

Lieferant: Apollo Scientific

Synonyme: TBAT, Tetrabutylammonium difluorotriphenylsilicate(IV)

PC905582-25G
APOSPC905582-25GEA 109 EUR
APOSPC905582-25G
Tetrabutylammonium difluorotriphenylsilicate 97%
Tetrabutylammonium difluorotriphenylsilicate
Formel: C₃₄H₅₁F₂NSi
Molecular Weight: 539,87 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00274218
CAS-Nummer: 163931-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR