4-Cyano-3,5-difluoropyridine 98%

Lieferant: Apollo Scientific

Synonyme: 3,5-Difluoroisonicotinonitrile

PC906325-5G
APOSPC906325-5GEA 66.7 EUR
APOSPC906325-5G
4-Cyano-3,5-difluoropyridine 98%
4-Cyano-3,5-difluoropyridine
Formel: C₆H₂F₂N₂
Molecular Weight: 140,093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176565
CAS-Nummer: 1214377-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR