3-Fluoro-5-methylpyridin-2-amine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-3-fluoro-5-methylpyridine

PC906342-1G PC906342-5G
APOSPC906342-1GEA 219 EUR
APOSPC906342-1G APOSPC906342-5G
3-Fluoro-5-methylpyridin-2-amine 95%
3-Fluoro-5-methylpyridin-2-amine
Formel: C₆H₇FN₂
Molecular Weight: 126.134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185333
CAS-Nummer: 1211590-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR