1-(Trifluormethyl)cyclopentancarbonsäure 95%

Lieferant: Apollo Scientific

PC906373-1G
APOSPC906373-1GEA 423 EUR
APOSPC906373-1G
1-(Trifluormethyl)cyclopentancarbonsäure 95%
1-(Trifluoromethyl)cyclopentanecarboxylic acid
Formel: C₇H₉F₃O₂
Molecular Weight: 182,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445821
CAS-Nummer: 277756-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR