3-[4-(Brommethyl)phenyl]-3-(trifluormethyl)-3H-diaziren 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(4-(Bromomethyl)phenyl)-3-(trifluoromethyl)-3H-diazirine, 3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
PC906453-1G
PC906453-200MG
APOSPC906453-1GEA
524
EUR
APOSPC906453-1G
APOSPC906453-200MG
3-[4-(Brommethyl)phenyl]-3-(trifluormethyl)-3H-diaziren 95+%
3-[4-(Brommethyl)phenyl]-3-(trifluormethyl)-3H-diaziren
Formel:
C₉H₆BrF₃N₂ Molecular Weight: 279,06 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD19442581 CAS-Nummer: 92367-11-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...