4,5-Difluorophthalonitrile 95%
Lieferant: Apollo Scientific
Synonyme:
1,2-Dicyano-4,5-difluorobenzene
PC906503-1G
PC906503-5G
APOSPC906503-1GEA
77
EUR
APOSPC906503-1G
APOSPC906503-5G
4,5-Difluorophthalonitrile 95%
4,5-Difluorophthalonitrile
Formel:
C₈H₂F₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061281 CAS-Nummer: 134450-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...