3-(Trifluoromethyl)quinoline 95%

Lieferant: Apollo Scientific

PC906529-1G PC906529-250MG PC906529-5G
APOSPC906529-1GEA 255 EUR
APOSPC906529-1G APOSPC906529-250MG APOSPC906529-5G
3-(Trifluoromethyl)quinoline 95%
3-(Trifluoromethyl)quinoline
Formel: C₁₀H₆F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD11007731
CAS-Nummer: 25199-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR