(2-Fluorophenyl)oxoacetic acid 95%

Lieferant: Apollo Scientific

PC906569-1G PC906569-5G
APOSPC906569-1GEA 319 EUR
APOSPC906569-1G APOSPC906569-5G
(2-Fluorophenyl)oxoacetic acid 95%
(2-Fluorophenyl)oxoacetic acid
Formel: C₈H₅FO₃
Molecular Weight: 168,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739153
CAS-Nummer: 79477-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR