5-Fluoropyrimidine-4,6-diol 95%
Lieferant: Apollo Scientific
PC907348-5G
APOSPC907348-5GEA
89.9
EUR
APOSPC907348-5G
5-Fluoropyrimidine-4,6-diol 95%
5-Fluoropyrimidine-4,6-diol
Formel:
C₄H₃FN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033260 CAS-Nummer: 106615-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...