2-Amino-2-(4-fluorophenyl)ethanol 97%
Lieferant: Apollo Scientific
Synonyme:
b-Amino-4-fluorobenzeneethanol
PC907360-1G
PC907360-250MG
APOSPC907360-1GEA
125
EUR
APOSPC907360-1G
APOSPC907360-250MG
2-Amino-2-(4-fluorophenyl)ethanol 97%
2-Amino-2-(4-fluorophenyl)ethanol
Formel:
C₈H₁₀FNO Molecular Weight: 155,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11213644 CAS-Nummer: 140373-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...