3-Fluor-4-hydroxybenzaldehyd

Lieferant: Apollo Scientific
Achtung

PC9083-25G PC9083-5G PC9083-100G
APOSPC9083-25GEA 148 EUR
APOSPC9083-25G APOSPC9083-5G APOSPC9083-100G
3-Fluor-4-hydroxybenzaldehyd
3-Fluoro-4-hydroxybenzaldehyde
Formel: C₇H₅FO₂
Molecular Weight: 140.11 g/mol
Schmelzpunkt: 121…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016626
CAS-Nummer: 405-05-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR