Pentafluorophenyl Benzoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzoic acid Pentafluorophenyl ester

PC908537-1G PC908537-5G
APOSPC908537-1GEA 34.8 EUR
APOSPC908537-1G APOSPC908537-5G
Pentafluorophenyl Benzoate 98%
Pentafluorophenyl Benzoate
Formel: C₁₃H₅F₅O₂
Molecular Weight: 288,17 g/mol
Schmelzpunkt: 73…74 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 1548-84-1
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR