3-Amino-6-fluor-1H-indazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Fluoro-1H-indazol-3-amine
PC908712-1G
PC908712-250MG
PC908712-5G
APOSPC908712-1GEA
60.9
EUR
APOSPC908712-1G
APOSPC908712-250MG
APOSPC908712-5G
3-Amino-6-fluor-1H-indazol 97%
3-Amino-6-fluoro-1H-indazole
Formel:
C₇H₆FN₃ Molecular Weight: 151,1400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739129 CAS-Nummer: 404827-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...