2-Chlor-5-fluorbenzotrifluorid 95%
Lieferant: Apollo Scientific
PC9088-25G
PC9088-5G
PC9088-1G
APOSPC9088-25GEA
79.9
EUR
APOSPC9088-25G
APOSPC9088-5G
APOSPC9088-1G
2-Chlor-5-fluorbenzotrifluorid 95%
2-Chlor-5-fluorbenzotrifluorid
Formel:
C₇H₃ClF₄ Molecular Weight: 198,5500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782058 CAS-Nummer: 89634-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...