Ethyl(4-fluorbenzoyl)acetat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-(4-Fluorophenyl)-3-oxopropionate, 3-(4-Fluorophenyl)-3-oxopropionic Acid Ethyl Ester, (4-Fluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-fluorophenyl)-3-oxopropanoate

PC9093-25G
APOSPC9093-25GEA 85.7 EUR
APOSPC9093-25G
Ethyl(4-fluorbenzoyl)acetat 97%
Ethyl (4-fluorobenzoyl)acetate
Formel: C₁₁H₁₁FO₃
Molecular Weight: 210,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093631
CAS-Nummer: 1999-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR