Methyl (4-bromo-3-fluorophenyl)acetate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(4-bromo-3-fluorophenyl)acetate

PC909748-1G
APOSPC909748-1GEA 48 EUR
APOSPC909748-1G
Methyl (4-bromo-3-fluorophenyl)acetate 95%
Methyl (4-bromo-3-fluorophenyl)acetate
Formel: C₉H₈BrFO₂
Molecular Weight: 247,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22418249
CAS-Nummer: 942282-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR