4-Fluor-3-iodbenzaldehyd

Lieferant: Apollo Scientific
Achtung

PC9099-1G PC9099-250MG PC9099-5G
APOSPC9099-1GEA 50.9 EUR
APOSPC9099-1G APOSPC9099-250MG APOSPC9099-5G
4-Fluor-3-iodbenzaldehyd
4-Fluoro-3-iodobenzaldehyde
Formel: C₇H₄OFI
Molecular Weight: 250,01 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08236850
CAS-Nummer: 227609-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR