4-(Difluoromethoxy)chlorobenzene 95%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-4-(difluoromethoxy)benzene

PC910104-25G PC910104-5G
APOSPC910104-25GEA 223 EUR
APOSPC910104-25G APOSPC910104-5G
4-(Difluoromethoxy)chlorobenzene 95%
4-(Difluoromethoxy)chlorobenzene
Formel: C₇H₅ClF₂O
Molecular Weight: 178,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09879256
CAS-Nummer: 81932-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR