4-Fluor-3-iodbenzonitril

Lieferant: Apollo Scientific
Achtung

PC9106-1G PC9106-5G PC9106-250MG
APOSPC9106-1GEA 59.6 EUR
APOSPC9106-1G APOSPC9106-5G APOSPC9106-250MG
4-Fluor-3-iodbenzonitril
4-Fluor-3-iodbenzonitril
Formel: C₇H₃FIN
Molecular Weight: 247,100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458123
CAS-Nummer: 159719-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR