3-Fluor-N-propylbenzylamin 98%
Lieferant: Apollo Scientific
Synonyme:
N-(3-Fluorobenzyl)propylamine
PC910859-1G
PC910859-5G
APOSPC910859-1GEA
219
EUR
APOSPC910859-1G
APOSPC910859-5G
3-Fluor-N-propylbenzylamin 98%
3-Fluor-N-propylbenzylamin
Formel:
C₁₀H₁₄FN Molecular Weight: 167,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09938496 CAS-Nummer: 90389-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...