3-(2-Fluorophenyl)pyrrolidine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 3-(2-Fluorophenyl)pyrrolidine

PC911081-100MG PC911081-1G PC911081-250MG
APOSPC911081-100MGEA 168 EUR
APOSPC911081-100MG APOSPC911081-1G APOSPC911081-250MG
3-(2-Fluorophenyl)pyrrolidine hydrochloride 95%
3-(2-Fluorophenyl)pyrrolidine hydrochloride
Formel: C₁₀H₁₃ClFN
Molecular Weight: 201.67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094757
CAS-Nummer: 885277-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR