(2,2,2-Trifluoro-1-pyridin-2-ylethyl)amine dihydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 2,2,2-Trifluoro-1-(pyridin-2-yl)ethanamine

PC911215-1G PC911215-250MG
APOSPC911215-1GEA 632 EUR
APOSPC911215-1G APOSPC911215-250MG
(2,2,2-Trifluoro-1-pyridin-2-ylethyl)amine dihydrochloride 95%
(2,2,2-Trifluoro-1-pyridin-2-ylethyl)amine dihydrochloride
Formel: C₇H₉Cl₂F₃N₂
Molecular Weight: 249.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16872039
CAS-Nummer: 503173-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR