[3-(2-Fluorophenoxy)propyl]amine 95%

Lieferant: Apollo Scientific

PC911359-1G PC911359-250MG
APOSPC911359-1GEA 574 EUR
APOSPC911359-1G APOSPC911359-250MG
[3-(2-Fluorophenoxy)propyl]amine 95%
[3-(2-Fluorophenoxy)propyl]amine
Formel: C₉H₁₂FNO
Molecular Weight: 169,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06800759
CAS-Nummer: 116735-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR