N,N-Diethyl chlorofluoroacetamide 97%

Lieferant: Apollo Scientific
Achtung

PC9117-25G PC9117-5G
APOSPC9117-25GEA 531 EUR
APOSPC9117-25G APOSPC9117-5G
N,N-Diethyl chlorofluoroacetamide 97%
N,N-Diethyl chlorofluoroacetamide
Formel: C₆H₁₁ClFNO
Siedepunkt: 113…115 °C/27mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD00101818
CAS-Nummer: 364-94-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR