3'-Chlor-2',6'-difluoracetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Chloro-2,6-difluorophenyl)ethanone
PC9122-5G
PC9122-1G
PC9122-25G
APOSPC9122-5GEA
102
EUR
APOSPC9122-5G
APOSPC9122-1G
APOSPC9122-25G
3'-Chlor-2',6'-difluoracetophenon 98%
3'-Chlor-2',6'-difluoracetophenon
Formel:
C₈H₅ClF₂O Molecular Weight: 190,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631329 CAS-Nummer: 177942-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...