3-Bromo-2-(trifluoromethyl)pyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9129-1G
PC9129-5G
APOSPC9129-1GEA
21.9
EUR
APOSPC9129-1G
APOSPC9129-5G
3-Bromo-2-(trifluoromethyl)pyridine 97%
3-Bromo-2-(trifluoromethyl)pyridine
Formel:
C6H3BrF3N Molecular Weight: 225,9939 g/mol Siedepunkt: 97…99 °C/26mmHg Schmelzpunkt: 40…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458124 CAS-Nummer: 590371-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...