5-Amino-2-fluorpyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9133-10G
PC9133-250G
PC9133-50G
APOSPC9133-10GEA
0
EUR
APOSPC9133-10G
APOSPC9133-250G
APOSPC9133-50G
5-Amino-2-fluorpyridin 97%
5-Amino-2-fluorpyridin
Formel:
C₅H₅FN₂ Schmelzpunkt: 86…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632180 CAS-Nummer: 1827-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...