5-(Trifluoromethyl)pyridine-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Trifluoromethyl)nicotinic acid

PC9138-1G PC9138-5G
APOSPC9138-1GEA 24.8 EUR
APOSPC9138-1G APOSPC9138-5G
5-(Trifluoromethyl)pyridine-3-carboxylic acid 97%
5-(Trifluoromethyl)pyridine-3-carboxylic acid
Formel: C₇H₄F₃NO₂
Molecular Weight: 191.11 g/mol
Schmelzpunkt: 184…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07774128
CAS-Nummer: 131747-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR