1-Fluor-4-(methylsulfonyl)-2-nitrobenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-methylsulphonylnitrobenzene, 1-Fluoro-4-(methylsulphonyl)-2-nitrobenzene

PC9145-25G PC9145-5G
APOSPC9145-25GEA 58.2 EUR
APOSPC9145-25G APOSPC9145-5G
1-Fluor-4-(methylsulfonyl)-2-nitrobenzol
1-Fluor-4-(methylsulfonyl)-2-nitrobenzol
Formel: C₇H₆FNO₄S
Molecular Weight: 219,19 g/mol
Schmelzpunkt: 148…157 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00025066
CAS-Nummer: 453-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR