4-Methyl-3-nitrobenzotrifluorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-2-nitro-4-(trifluoromethyl)benzene

PC9163-1G PC9163-5G PC9163-25G
APOSPC9163-1GEA 21.9 EUR
APOSPC9163-1G APOSPC9163-5G APOSPC9163-25G
4-Methyl-3-nitrobenzotrifluorid 97%
4-Methyl-3-nitrobenzotrifluorid
Formel: C₈H₆F₃NO₂
Molecular Weight: 205.1300 g/mol
Siedepunkt: 76…78 °C/8mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00272402
CAS-Nummer: 65754-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR