2-Chlor-5-fluorbenzylamin 97%

Lieferant: Apollo Scientific
Gefahr

PC9177-5G PC9177-1G PC9177-25G
APOSPC9177-5GEA 116 EUR
APOSPC9177-5G APOSPC9177-1G APOSPC9177-25G
2-Chlor-5-fluorbenzylamin 97%
2-Chlor-5-fluorbenzylamin
Formel: C₇H₇ClFN
Molecular Weight: 159,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213626
CAS-Nummer: 202522-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR