1,2,2-Trifluoro-2-hydroxy-1-(trifluoromethyl)ethanesulphonic acid sultone 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3,4,4-Trifluoro-3-(trifluoromethyl)-1,2-oxathietane 2,2-dioxide
PC9181-25G
PC9181-5G
APOSPC9181-25GEA
522
EUR
APOSPC9181-25G
APOSPC9181-5G
1,2,2-Trifluoro-2-hydroxy-1-(trifluoromethyl)ethanesulphonic acid sultone 95%
1,2,2-Trifluoro-2-hydroxy-1-(trifluoromethyl)ethanesulphonic acid sultone
Formel:
C₃F₆O₃S Molecular Weight: 230,09 g/mol Siedepunkt: 46,5 °C Dichte: 1,667 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082475 CAS-Nummer: 773-15-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...