3-Amino-2-fluorpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluorpyridin-3-amin, 2-Fluor-3-pyridylamin
PC9189-1G
PC9189-5G
APOSPC9189-1GEA
21.9
EUR
APOSPC9189-1G
APOSPC9189-5G
3-Amino-2-fluorpyridin
3-Amino-2-fluorpyridin
Formel:
C₅H₅FN₂ Molecular Weight: 112,11 g/mol Dichte: 1,212 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095248 CAS-Nummer: 1597-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...