3-Amino-α,α,α-trifluor-p-toluylsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4-(trifluoromethyl)benzoic acid

PC9194-10G PC9194-1G PC9194-5G
APOSPC9194-10GEA 132 EUR
APOSPC9194-10G APOSPC9194-1G APOSPC9194-5G
3-Amino-α,α,α-trifluor-p-toluylsäure 97%
3-Amino-α,α,α-trifluor-p-toluylsäure
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,1340 g/mol
Schmelzpunkt: 198,6…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784307
CAS-Nummer: 125483-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR