2-Iodo-5-trifluoromethylpyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Iodo-5-(trifluoromethyl)pyridine

PC9199-25G PC9199-5G
APOSPC9199-25GEA 134 EUR
APOSPC9199-25G APOSPC9199-5G
2-Iodo-5-trifluoromethylpyridine 98%
2-Iodo-5-trifluoromethylpyridine
Formel: C₆H₃F₃IN
Molecular Weight: 272.99 g/mol
Schmelzpunkt: 79…83 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08741351
CAS-Nummer: 100366-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR