Ethyl-3-{[3-(trifluoromethyl)benzoyl]amino}-1H-indole-2-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-[3-(trifluoromethyl)benzamido]-1H-indole-2-carboxylate

PC9211-1G PC9211-500MG
APOSPC9211-1GEA 488 EUR
APOSPC9211-1G APOSPC9211-500MG
Ethyl-3-{[3-(trifluoromethyl)benzoyl]amino}-1H-indole-2-carboxylate 97%
Ethyl-3-{[3-(trifluoromethyl)benzoyl]amino}-1H-indole-2-carboxylate
Formel: C₁₉H₁₅F₃N₂O₃
Molecular Weight: 376,34 g/mol
Schmelzpunkt: 212…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00794680
CAS-Nummer: 338968-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR