5-Amino-2-(trifluormethyl)benzimidazol 96%

Lieferant: Apollo Scientific
Gefahr

PC9213-1G PC9213-5G
APOSPC9213-1GEA 53.8 EUR
APOSPC9213-1G APOSPC9213-5G
5-Amino-2-(trifluormethyl)benzimidazol 96%
5-Amino-2-(trifluoromethyl)benzimidazole
Formel: C₈H₆F₃N₃
Molecular Weight: 201,15 g/mol
Schmelzpunkt: 185…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632182
CAS-Nummer: 3671-66-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR