2-Fluor-5-(trifluormethyl)zimtsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC9243-1G
PC9243-5G
APOSPC9243-1GEA
21.9
EUR
APOSPC9243-1G
APOSPC9243-5G
2-Fluor-5-(trifluormethyl)zimtsäure
2-Fluor-5-(trifluormethyl)zimtsäure
Formel:
C₁₀H₆F₄O₂ Molecular Weight: 234.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236293 CAS-Nummer: 247113-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...