1,1,2,3,3,3-Hexafluorpropyldichlormethylether 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC9252-25G
PC9252-5G
APOSPC9252-25GEA
143
EUR
APOSPC9252-25G
APOSPC9252-5G
1,1,2,3,3,3-Hexafluorpropyldichlormethylether 98%
1,1,2,3,3,3-Hexafluorpropyldichlormethylether
Formel:
C₄H₂Cl₂F₆O Siedepunkt: 98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094547 CAS-Nummer: 56860-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...