Methyl-3-[2-nitro-4-(trifluoromethyl)phenoxy]thiophene-2-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9258-1G
PC9258-10G
APOSPC9258-1GEA
102
EUR
APOSPC9258-1G
APOSPC9258-10G
Methyl-3-[2-nitro-4-(trifluoromethyl)phenoxy]thiophene-2-carboxylate 97%
Methyl-3-[2-nitro-4-(trifluoromethyl)phenoxy]thiophene-2-carboxylate
Molecular Weight:
306.24 g/mol Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00112557 CAS-Nummer: 91041-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...