3-Brom-5-fluorphenylessigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3-Bromo-5-fluorophenyl)acetic acid
PC9278-1G
PC9278-5G
PC9278-25G
APOSPC9278-1GEA
52.2
EUR
APOSPC9278-1G
APOSPC9278-5G
APOSPC9278-25G
3-Brom-5-fluorphenylessigsäure 98%
3-Brom-5-fluorphenylessigsäure
Formel:
C₈H₆BrFO₂ Molecular Weight: 233.0300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258961 CAS-Nummer: 202000-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...